Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151382.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151382.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151382.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151382.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151382.hg.1 |
TC0900007601.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900151381.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151381.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151381.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151381.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151381.hg.1 |
TC0900007601.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007601.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151383.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151383.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151383.hg.1 |
ENST00000455609.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151383.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151383.hg.1 |
TC0900007601.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900151384.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151384.hg.1 |
NR_121183.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151384.hg.1 |
TC0900007601.hg.1 |
Clariom D TC |