Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
ENST00000435037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
ENST00000435037.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165454.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165454.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165454.hg.1 |
TC1300009818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300165452.hg.1 |
ENST00000435037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165452.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165452.hg.1 |
ENST00000435037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165452.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165452.hg.1 |
TC1300009818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300165457.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165457.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165457.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165457.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165457.hg.1 |
TC1300009818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300165453.hg.1 |
ENST00000435037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165453.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165453.hg.1 |
ENST00000435037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165453.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165453.hg.1 |
TC1300009818.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
ENST00000435037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
ENST00000435037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_046516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009818.hg.1 |
NR_120398.1 |
Refseq |