Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008465.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008465.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008465.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008465.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008465.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008465.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008465.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008465.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160284.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160284.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160284.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160284.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160284.hg.1 |
TC0300008465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008465.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008465.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008465.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008465.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008465.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008465.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008465.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008465.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160285.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160285.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160285.hg.1 |
ENST00000470790.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300160285.hg.1 |
NR_046574.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300160285.hg.1 |
TC0300008465.hg.1 |
Clariom D TC |