Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010026.hg.1 |
ENST00000451712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010026.hg.1 |
ENST00000451712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168471.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168471.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168471.hg.1 |
TC0600010026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600168467.hg.1 |
ENST00000451712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168467.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168467.hg.1 |
ENST00000451712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168467.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168467.hg.1 |
TC0600010026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600168469.hg.1 |
ENST00000451712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168469.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168469.hg.1 |
ENST00000451712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168469.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168469.hg.1 |
TC0600010026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600010026.hg.1 |
ENST00000451712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010026.hg.1 |
ENST00000451712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010026.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168466.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168466.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168466.hg.1 |
TC0600010026.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600168470.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168470.hg.1 |
NR_102425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168470.hg.1 |
TC0600010026.hg.1 |
Clariom D TC |