Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000430181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000430181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000430181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149881.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149881.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149881.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100149881.hg.1 |
TC2100007370.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100149881.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149881.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100149881.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000430181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000430181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000430181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
ENST00000451035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007370.hg.1 |
NR_103707.1 |
Refseq |