Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000618159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000618159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000618159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000618159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000614132.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000618159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000618159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000618159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000618159.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007720.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
ENST00000610434.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
ENST00000613870.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
ENST00000619426.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
ENST00000620309.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
NM_002795.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
NR_104194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
NR_104195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700155701.hg.1 |
TC1700007720.hg.1 |
Clariom D TC |