Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001604.hg.1 |
TC0Y000161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001604.hg.1 |
ENST00000357871.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001604.hg.1 |
ENST00000382963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001604.hg.1 |
ENST00000382965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001604.hg.1 |
ENST00000382966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001604.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001606.hg.1 |
TC0Y000161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001606.hg.1 |
ENST00000357871.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001606.hg.1 |
ENST00000382966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001606.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001608.hg.1 |
TC0Y000161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001608.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001600.hg.1 |
TC0Y000161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001600.hg.1 |
ENST00000382963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001600.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001601.hg.1 |
TC0Y000161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001601.hg.1 |
ENST00000357871.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001601.hg.1 |
ENST00000382963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001601.hg.1 |
ENST00000382965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001601.hg.1 |
ENST00000382966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001601.hg.1 |
ENST00000493160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001601.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001602.hg.1 |
TC0Y000161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001602.hg.1 |
ENST00000382963.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001602.hg.1 |
ENST00000493160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0Y001602.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000382965.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147629.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147629.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147629.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147629.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147629.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147629.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147629.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147629.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147629.hg.1 |
TC0Y00007060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0Y00147630.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147630.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147630.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147630.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147630.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147630.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147630.hg.1 |
TC0Y00007060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0Y00147624.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147624.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147624.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147624.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147624.hg.1 |
TC0Y00007060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000382965.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000382965.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147625.hg.1 |
TC0Y00007060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0Y00147626.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147626.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147626.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147626.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147626.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00147626.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147626.hg.1 |
TC0Y00007060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0Y00147632.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147632.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00147632.hg.1 |
TC0Y00007060.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000357871.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000382963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000382965.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000382966.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
ENST00000493160.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007060.hg.1 |
NR_033667.1 |
Refseq |