Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011279.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011279.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011279.hg.1 |
TC22000647.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011283.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011283.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011283.hg.1 |
TC22000647.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011280.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011280.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011280.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011280.hg.1 |
TC22000647.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200160406.hg.1 |
TC2200008425.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160406.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160406.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160406.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160406.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160406.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160406.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160406.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160406.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
ENST00000403987.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008425.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160409.hg.1 |
TC2200008425.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160409.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160409.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160409.hg.1 |
NM_002309.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160409.hg.1 |
ENST00000249075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160409.hg.1 |
NM_001257135.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160409.hg.1 |
NM_002309.4 |
Refseq |