Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002412.4 | NP_002403.2 | 4 | 440 | 76 | |
XM_005252682.2 | XP_005252739.1 | 4 | 477 | 76 | ||
XM_017016275.1 | XP_016871764.1 | 3 | 263 | 76 | ||
GenBank mRNA | AK293593.1 | - | 4 | 410 | 76 | |
BC000824.1 | - | 4 | 368 | 3454207 | 76 | |
BI520029.1 | - | 4 | 422 | 76 | ||
BT006714.1 | - | 3 | 315 | 76 | ||
DC387686.1 | - | 4 | 434 | 76 | ||
DC416206.1 | - | 4 | 440 | 76 | ||
M29971.1 | - | 4 | 411 | 76 | ||
M31767.1 | - | 3 | 337 | 76 | ||
M60761.1 | - | 3 | 330 | 76 | ||
X54228.1 | - | 3 | 355 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |