Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000334662.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000334662.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000334662.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000334662.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000616689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000616689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000616689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
ENST00000616689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007375.hg.1 |
NM_198695.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002791.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002791.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002791.hg.1 |
TC21000237.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000397907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000397907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000397907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000397907.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000618832.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000619249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000619249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000619249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
ENST00000619249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198697.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002790.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002790.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21002790.hg.1 |
TC21000237.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100158415.hg.1 |
TC2100008530.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100158415.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158415.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158415.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158415.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158415.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158415.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158413.hg.1 |
TC2100008530.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100158413.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158413.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158413.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158413.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158417.hg.1 |
TC2100008530.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100158417.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158417.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158417.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158417.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000334662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000334662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000334662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000334662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000609664.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000616689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000616689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000616689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
ENST00000616689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198689.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198695.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198695.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198695.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007375.hg.1 |
NM_198695.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158414.hg.1 |
TC2100008530.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100158414.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158414.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158414.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100158414.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158414.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100158414.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000334670.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000397907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000397907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000397907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000397907.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000400365.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000618832.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000619249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000619249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000619249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
ENST00000619249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198692.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198697.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008530.hg.1 |
NM_198699.1 |
Refseq |