Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098209.1 | NP_001091679.1 | 9 - 10 | 1796 | 134 | |
NM_001098210.1 | NP_001091680.1 | 9 - 10 | 1796 | 134 | ||
NM_001904.3 | NP_001895.1 | 9 - 10 | 1796 | 134 | ||
XM_005264886.2 | XP_005264943.1 | 9 - 10 | 1810 | 134 | ||
XM_006712983.1 | XP_006713046.1 | 9 - 10 | 1708 | 134 | ||
XM_006712984.1 | XP_006713047.1 | 9 - 10 | 1708 | 134 | ||
XM_006712985.1 | XP_006713048.1 | 9 - 10 | 1770 | 134 | ||
XM_017005738.1 | XP_016861227.1 | 9 - 10 | 1810 | 134 | ||
GenBank mRNA | AB062292.1 | - | 4 - 5 | 759 | 134 | |
AB451264.1 | - | 7 - 8 | 1528 | 134 | ||
AB451392.1 | - | 7 - 8 | 1528 | 134 | ||
AK289932.1 | - | 10 - 11 | 1684 | 134 | ||
AK294778.1 | - | 10 - 11 | 1909 | 134 | ||
AK296790.1 | - | 9 - 10 | 1616 | 134 | ||
AK299954.1 | - | 9 - 10 | 1478 | 134 | ||
AK314776.1 | - | 9 - 10 | 1742 | 134 | ||
BC058926.1 | - | 9 - 10 | 1738 | 6151332 | 134 | |
X87838.1 | - | 9 - 10 | 1742 | 134 | ||
Z19054.1 | - | 9 - 10 | 1728 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |