Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283089.1 | NP_001270018.1 | 7 - 8 | 680 | 96 | |
XM_015450547.1 | XP_015306033.1 | 7 - 8 | 1051 | 95 | ||
XM_015450548.1 | XP_015306034.1 | 7 - 8 | 758 | 95 | ||
XM_015450549.1 | XP_015306035.1 | 7 - 8 | 886 | 95 | ||
XM_015450550.1 | XP_015306036.1 | 7 - 8 | 807 | 95 | ||
XM_015450552.1 | XP_015306038.1 | 6 - 7 | 553 | 95 | ||
GenBank mRNA | AB169509.1 | - | 7 - 8 | 598 | 95 | |
AB171036.1 | - | 7 - 8 | 680 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |