Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANEAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543924.2 | XP_005543981.1 | 2 - 3 | 357 | 73 | |
XM_005543925.2 | XP_005543982.2 | 3 - 4 | 614 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |