Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015449302.1 | XP_015304788.1 | 9 - 10 | 3003 | 68 | |
XM_015449307.1 | XP_015304793.1 | 9 - 10 | 3003 | 68 | ||
XM_015449309.1 | XP_015304795.1 | 8 - 9 | 2861 | 68 | ||
XM_015449313.1 | XP_015304799.1 | 9 - 10 | 2270 | 68 | ||
XM_015449318.1 | XP_015304804.1 | 9 - 10 | 3009 | 68 | ||
XM_015449320.1 | XP_015304806.1 | 9 - 10 | 2271 | 68 | ||
XM_015449323.1 | XP_015304809.1 | 9 - 10 | 3004 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |