Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566756.2 | XP_005566813.1 | 14 - 15 | 4532 | 60 | |
XM_005566758.2 | XP_005566815.1 | 14 - 15 | 2055 | 60 | ||
XM_005566760.2 | XP_005566817.1 | 14 - 15 | 4532 | 60 | ||
XM_005566762.2 | XP_005566819.1 | 13 - 14 | 4314 | 60 | ||
XM_005566763.2 | XP_005566820.1 | 13 - 14 | 4308 | 60 | ||
XM_015456882.1 | XP_015312368.1 | 13 - 14 | 4260 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |