Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284843.1 | NP_001271772.1 | 3 - 4 | 338 | 88 | |
XM_005585079.2 | XP_005585136.1 | 3 - 4 | 344 | 88 | ||
XM_005585080.2 | XP_005585137.1 | 3 - 4 | 341 | 88 | ||
XM_005585081.2 | XP_005585138.1 | 2 - 3 | 515 | 88 | ||
XM_015437827.1 | XP_015293313.1 | 3 - 4 | 341 | 88 | ||
GenBank mRNA | AB179020.1 | - | 3 - 4 | 338 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |