Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285221.1 | NP_001272150.1 | 3 - 4 | 1090 | 91 | |
XM_015454799.1 | XP_015310285.1 | 3 - 4 | 1106 | 91 | ||
XM_015454800.1 | XP_015310286.1 | 3 - 4 | 1426 | 91 | ||
XM_015454801.1 | XP_015310287.1 | 2 - 3 | 2595 | 91 | ||
XM_015454802.1 | XP_015310288.1 | 3 - 4 | 2349 | 91 | ||
XM_015454804.1 | XP_015310290.1 | 3 - 4 | 1015 | 91 | ||
GenBank mRNA | AB171987.1 | - | 3 - 4 | 1090 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |