Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LANCL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593275.2 | XP_005593332.1 | 2 - 3 | 1020 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |