Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005580147.2 | XP_005580204.1 | 1 - 2 | 2226 | 81 | |
XM_005580148.2 | XP_005580205.1 | 2 - 3 | 1343 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |