Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581569.2 | XP_005581626.1 | 7 - 8 | 1166 | 103 | |
XM_005581570.2 | XP_005581627.1 | 7 - 8 | 1057 | 103 | ||
XM_005581571.2 | XP_005581628.1 | 7 - 8 | 1167 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |