Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591416.2 | XP_005591473.1 | 3 - 4 | 646 | 133 | |
XM_005591417.2 | XP_005591474.1 | 3 - 4 | 476 | 133 | ||
XM_005591418.2 | XP_005591475.1 | 4 - 5 | 624 | 133 | ||
XM_005591419.2 | XP_005591476.1 | 3 - 4 | 624 | 133 | ||
XM_015442271.1 | XP_015297757.1 | 3 - 4 | 351 | 133 | ||
XM_015442272.1 | XP_015297758.1 | 3 - 4 | 873 | 133 | ||
XM_015442273.1 | XP_015297759.1 | 3 - 4 | 267 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |