Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015452481.1 | XP_015307967.1 | 5 - 6 | 2025 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |