Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005556398.2 | XP_005556455.1 | 11 - 12 | 1062 | 117 | |
XM_015451081.1 | XP_015306567.1 | 11 - 12 | 954 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |