Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015439339.1 | XP_015294825.1 | 6 - 7 | 1121 | 100 | |
XM_015439340.1 | XP_015294826.1 | 6 - 7 | 1228 | 100 | ||
XM_015439341.1 | XP_015294827.1 | 6 - 7 | 2309 | 100 | ||
XM_015439342.1 | XP_015294828.1 | 4 - 5 | 922 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |