Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579724.2 | XP_005579781.1 | 2 - 3 | 115 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |