Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005567148.2 | XP_005567205.1 | 2 - 3 | 187 | 62 | |
XM_005567149.2 | XP_005567206.1 | 2 - 3 | 187 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |