Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSPRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005581016.1 | XP_005581073.1 | 2 - 3 | 357 | 103 | |
XM_005581017.1 | XP_005581074.1 | 2 - 3 | 346 | 103 | ||
XM_015436837.1 | XP_015292323.1 | 2 - 3 | 357 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |