Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589334.2 | XP_005589391.1 | 2 - 3 | 969 | 71 | |
XM_005589335.2 | XP_005589392.1 | 2 - 3 | 969 | 71 | ||
XM_005589336.2 | XP_005589393.1 | 2 - 3 | 966 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |