Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285209.1 | NP_001272138.1 | 3 - 4 | 347 | 74 | |
XM_005563160.2 | XP_005563217.1 | 3 - 4 | 353 | 74 | ||
XM_015454376.1 | XP_015309862.1 | 3 - 4 | 353 | 74 | ||
XM_015454377.1 | XP_015309863.1 | 3 - 4 | 353 | 74 | ||
GenBank mRNA | AB173107.1 | - | 3 - 4 | 347 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |