Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560149.2 | XP_005560206.1 | 4 - 5 | 557 | 84 | |
XM_015453120.1 | XP_015308606.1 | 4 - 5 | 1035 | 84 | ||
XM_015453121.1 | XP_015308607.1 | 5 - 6 | 714 | 84 | ||
XM_015453122.1 | XP_015308608.1 | 5 - 6 | 1974 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |