Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571702.2 | XP_005571759.1 | 7 | 6022 | 133 | |
XM_005571703.2 | XP_005571760.1 | 7 | 6022 | 133 | ||
XM_005571704.2 | XP_005571761.1 | 7 | 6145 | 133 | ||
XM_005571708.2 | XP_005571765.1 | 6 | 5969 | 133 | ||
XM_005571709.2 | XP_005571766.1 | 7 | 5591 | 133 | ||
XM_005571710.2 | XP_005571767.1 | 6 | 5331 | 133 | ||
XM_005571711.2 | XP_005571768.1 | 4 | 8031 | 133 | ||
XM_005571712.2 | XP_005571769.1 | 4 | 6121 | 133 | ||
XM_015431315.1 | XP_015286801.1 | 5 | 5726 | 133 | ||
XM_015431316.1 | XP_015286802.1 | 5 | 5294 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |