Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283603.1 | NP_001270532.1 | 3 - 4 | 468 | 84 | |
XM_005562595.2 | XP_005562652.1 | 3 - 4 | 1446 | 84 | ||
XM_005562601.2 | XP_005562658.1 | 3 - 4 | 2509 | 84 | ||
XM_005562602.2 | XP_005562659.1 | 4 - 5 | 2575 | 84 | ||
XM_015454378.1 | XP_015309864.1 | 3 - 4 | 1699 | 84 | ||
XM_015454379.1 | XP_015309865.1 | 2 - 3 | 1639 | 84 | ||
XM_015454382.1 | XP_015309868.1 | 5 - 6 | 3739 | 84 | ||
XM_015454384.1 | XP_015309870.1 | 3 - 4 | 1471 | 84 | ||
XM_015454385.1 | XP_015309871.1 | 3 - 4 | 3528 | 84 | ||
XM_015454386.1 | XP_015309872.1 | 3 - 4 | 1464 | 84 | ||
XM_015454387.1 | XP_015309873.1 | 4 - 5 | 3601 | 84 | ||
XM_015454388.1 | XP_015309874.1 | 2 - 3 | 859 | 84 | ||
XM_015454389.1 | XP_015309875.1 | 2 - 3 | 2737 | 84 | ||
XM_015454390.1 | XP_015309876.1 | 3 - 4 | 1448 | 84 | ||
XM_015454391.1 | XP_015309877.1 | 3 - 4 | 1445 | 84 | ||
XM_015454392.1 | XP_015309878.1 | 3 - 4 | 1446 | 84 | ||
GenBank mRNA | AB220441.1 | - | 3 - 4 | 468 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |