Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576318.2 | XP_005576375.1 | 3 - 4 | 1166 | 88 | |
XM_005576319.2 | XP_005576376.1 | 3 - 4 | 1164 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |