Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285192.1 | NP_001272121.1 | 4 - 5 | 597 | 68 | |
GenBank mRNA | AB000823.1 | - | 4 - 5 | 597 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Inventoried Probe spans exons |