Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287310.1 | NP_001274239.1 | 1 - 2 | 408 | 118 | |
XM_005542952.2 | XP_005543009.1 | 2 - 3 | 550 | 118 | ||
XM_005542953.2 | XP_005543010.1 | 2 - 3 | 265 | 118 | ||
XM_015452345.1 | XP_015307831.1 | 2 - 3 | 495 | 118 | ||
GenBank mRNA | AB173150.1 | - | 1 - 2 | 408 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |