Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CISD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583979.2 | XP_005584036.1 | 3 - 4 | 428 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |