Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005593433.2 | XP_005593490.1 | 2 - 3 | 330 | 93 | |
XM_005593434.2 | XP_005593491.1 | 2 - 3 | 362 | 93 | ||
XM_005593435.2 | XP_005593492.1 | 2 - 3 | 342 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |