Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005542420.1 | XP_005542477.1 | 3 - 4 | 2327 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |