Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIKESHI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005579283.2 | XP_005579340.1 | 2 - 3 | 505 | 80 | |
XM_015435583.1 | XP_015291069.1 | 3 - 4 | 512 | 80 | ||
XM_015435584.1 | XP_015291070.1 | 3 - 4 | 645 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |