Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583207.1 | XP_005583264.1 | 3 - 4 | 507 | 121 | |
XM_005583208.2 | XP_005583265.1 | 2 - 3 | 271 | 121 | ||
XM_005583209.2 | XP_005583266.1 | 3 - 4 | 307 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |