Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015434082.1 | XP_015289568.1 | 1 - 2 | 588 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |