Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566739.2 | XP_005566796.1 | 1 - 2 | 519 | 169 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |