Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553718.2 | XP_005553775.1 | 1 - 2 | 97 | 128 | |
XM_005553723.1 | XP_005553780.1 | 1 - 2 | 99 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |