Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005545284.2 | XP_005545341.1 | 3 | 802 | 78 | |
XM_005545285.2 | XP_005545342.1 | 3 | 1138 | 78 | ||
XM_015444868.1 | XP_015300354.1 | 4 | 2480 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |