Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283182.1 | NP_001270111.1 | 2 - 3 | 731 | 79 | |
XM_015440188.1 | XP_015295674.1 | 3 - 4 | 441 | 79 | ||
GenBank mRNA | AB168527.1 | - | 2 - 3 | 731 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |