Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005553544.2 | XP_005553601.1 | 2 - 3 | 583 | 132 | |
XM_005553545.2 | XP_005553602.1 | 1 - 2 | 673 | 132 | ||
XM_015449627.1 | XP_015305113.1 | 1 - 2 | 673 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Probe spans exons |