Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005566851.2 | XP_005566908.1 | 7 - 8 | 748 | 131 | |
XM_015456988.1 | XP_015312474.1 | 7 - 8 | 894 | 131 | ||
XM_015456989.1 | XP_015312475.1 | 7 - 8 | 888 | 131 | ||
XM_015456990.1 | XP_015312476.1 | 8 - 9 | 975 | 131 | ||
XM_015456992.1 | XP_015312478.1 | 8 - 9 | 875 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |