Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289963.1 | NP_001276892.1 | 5 - 6 | 412 | 82 | |
XM_015452394.1 | XP_015307880.1 | 5 - 6 | 673 | 82 | ||
GenBank mRNA | AF200709.1 | - | 5 - 6 | 360 | 82 | |
KC734501.1 | - | 5 - 6 | 412 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Inventoried Probe spans exons |