Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102127891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594153.2 | XP_005594210.1 | 1 - 2 | 150 | 62 | |
XM_005594154.2 | XP_005594211.1 | 1 - 2 | 150 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |