Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102127891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005594152.2 | XP_005594209.1 | 2 - 3 | 215 | 90 | |
XM_005594154.2 | XP_005594211.1 | 2 - 3 | 233 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |